634395 Aldrich

Zirconium(IV) silicate

nanopowder, <100 nm particle size (BET), 98.5% trace metals basis

DOWNLOAD MSDS (PDF)

Synonym: Zirconium(IV) silicon oxide

Description

Features and Benefits

Often cited as a potential alternative gate dielectric to silica.1

Packaging

25, 100 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Zirconium(IV) oxide

nanopowder, <100 nm particle size (TEM)

Zirconium(IV) silicate

−325 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?