Linear Formula TiC0.7N0.3
Molecular Weight 604.77
MDL number MFCD01868685
| Related Categories | 22: Ti, Carbon, Ceramics, Ceramics by Element, Materials Science, |
| assay | ≥97% trace metals basis |
| form | nanopowder |
| particle size | <150 nm |
| density | 5.02 g/mL at 25 °C(lit.) |
25 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?