• USA Home
  • 63753 - O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol 20′000

EMAIL THIS PAGE TO A FRIEND
63753 Aldrich

O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol 20′000

Synonym: ‘Mercaptopolyethylene glycol 20′000 monomethyl ether’

Purchase

Description

Packaging

250 mg in poly tube

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?