CAS Number 62882-08-0
Empirical Formula (Hill Notation) C9H6N2O3
Molecular Weight 190.16
MDL number MFCD08276758
PubChem Substance ID 24882958
| Related Categories | Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Oxazoles |
| assay | 97% |
1 g in glass bottle
Reactant involved in synthesis of carbon-substituted iminium salts and investigations of its synthetic applications1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?