• USA Home
  • 652512 - Carbon nanotube, single-walled

EMAIL THIS PAGE TO A FRIEND
652512 Aldrich

Carbon nanotube, single-walled Green Alternative

short, 90% carbon basis, O.D. × L 1-2 nm × 0.5-2 μm

Synonym: SWCNT, SWNT, Single wall carbon nanotube

Purchase

Properties

Related Categories Alternative Energy, Carbon Nanomaterials, Carbon Nanotubes, Carbon Nanotubes and Fullerenes, Chemical Synthesis,
assay   90% carbon basis
composition   carbon, 90%
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
O.D. × L   1-2 nm × 0.5-2 μm
surface area   spec. surface area 480 m2/g
mp   3652-3697 °C(lit.)
density   1.7-1.9 g/cm3 at 25 °C(lit.)
bulk density   0.22‑0.27 g/mL

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?