EMAIL THIS PAGE TO A FRIEND
65691 Aldrich

Luviquat HOLD

~20% active ingredients in H2O

Synonym: Poly[(3-methyl-1-vinylimidazolium methyl sulfate)-co-(1-vinylcaprolactam)-co-(1-vinylpyrrolidone)], Polyquaternium 46

Purchase

Properties

Related Categories Biochemicals and Reagents, Cationic Detergents, Detergents, Detergents A to M
description   cationic
concentration   ~20% active ingredients in H2O
refractive index   n20/D 1.371
pH   5.0-8.0 (10% in H2O, active ingredients)
density   1.04 g/mL at 20 °C

Description

Packaging

1 L in poly bottle

5 L in poly drum

Legal Information

Luviquat is a trademark of BASF SE

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?