EMAIL THIS PAGE TO A FRIEND
661643 Aldrich

Deuterium oxide

99.99 atom % D (for D2O), contains 1% DSS-d6

Synonym: Heavy water, Water-d2

  • CAS Number 7789-20-0

  • Empirical Formula (Hill Notation) D2O

  • Molecular Weight 20.03

  •  MDL number MFCD00044636

Purchase

Properties

Related Categories Alphabetical Listings, D, Stable Isotopes More...
InChI Key   XLYOFNOQVPJJNP-ZSJDYOACSA-N
isotopic purity   99.99 atom % D (for D2O)
contains   1% DSS-d6
refractive index   n20/D 1.328(lit.)
bp   101.4 °C(lit.)
mp   3.8 °C(lit.)
density   1.107 g/mL at 25 °C(lit.)

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Price and Availability

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.

Stable Isotope
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ZC0230000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?