• USA Home
  • 661961 - 3-Bromo-5-butoxyphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
661961 Aldrich

3-Bromo-5-butoxyphenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Disubstituted Aryl Boronic Acids,
InChI Key   OWWZXOTWNAQSTF-UHFFFAOYSA-N
mp   116-123 °C

Description

Other Notes

Contains varying amounts of anhydride

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?