• USA Home
  • 662526 - 2,3-Dichlorothiophenol

EMAIL THIS PAGE TO A FRIEND
662526 Aldrich

2,3-Dichlorothiophenol

97%

Synonym: 2,3-Dichlorobenzenethiol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
InChI Key   QGRKONUHHGBHRB-UHFFFAOYSA-N
assay   97%
mp   54-60 °C

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?