• USA Home
  • 663352 - Bis(propyltetramethylcyclopentadienyl)strontium(II) DME Adduct

EMAIL THIS PAGE TO A FRIEND
663352 Aldrich

Bis(propyltetramethylcyclopentadienyl)strontium(II) DME Adduct

  • Empirical Formula (Hill Notation) C28H48O2Sr

  • Molecular Weight 504.30

  •  MDL number MFCD00269844

  •  PubChem Substance ID 329760111

Purchase

Properties

Related Categories CVD and ALD Precursors by Metal, Materials Science, Micro/NanoElectronics, Strontium, Vapor Deposition Precursors More...
InChI Key   PIRUAPSZTGIKQF-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 3 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
129.2 °F
Flash Point(C) 
54 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?