CAS Number 127543-80-0
Empirical Formula (Hill Notation) C11H9BrMg
Molecular Weight 245.40
MDL number MFCD02260227
PubChem Substance ID 24884665
| Related Categories | Alkyl, Chemical Synthesis, Grignard Reagents, Organometallic Reagents |
| concentration | 0.25 M in diethyl ether |
| density | 0.776 g/mL at 25 °C |
100 mL in Sure/Seal™
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?