• USA Home
  • 666106 - 3-(4,5-Dihydro-1H-imidazol-2-yl)phenol

EMAIL THIS PAGE TO A FRIEND
666106 Aldrich

3-(4,5-Dihydro-1H-imidazol-2-yl)phenol

97%

Purchase

Properties

Related Categories Building Blocks, C9 to C30+, Chemical Synthesis, Heterocyclic Building Blocks, Imidazoles More...
InChI Key   YMULJMVKZKHEEH-UHFFFAOYSA-N
assay   97%
mp   197-201 °C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?