• USA Home
  • 666157 - 3-Formyl-2-methoxyphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
666157 Aldrich

3-Formyl-2-methoxyphenylboronic acid

≥95%

Purchase

Properties

Related Categories Aldehydes, Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Building Blocks,
InChI Key   DUROSIJIMLRFHR-UHFFFAOYSA-N
assay   ≥95%
mp   124-128 °C
storage temp.   2-8°C

Description

Other Notes

May contain varying amounts of anhydride

Packaging

1, 5 g in glass bottle

Application

Reactant for:
• Preparation of formyl-substituted aryl MIDA boronates
• Preparation of indolylamino phenylthienopyridine carbonitrile PKCθ inhibitors

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?