• USA Home
  • 666823 - 2-[(2′,6′-Diisopropylphenoxy)methyl]phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
666823 Aldrich

2-[(2′,6′-Diisopropylphenoxy)methyl]phenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   HPQFMTHWDXHLCU-UHFFFAOYSA-N
mp   109-116 °C

Description

General description

May contain varying amounts of anhydride

Packaging

2, 10 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?