• USA Home
  • 666890 - 6-Bromo-2-fluoro-3-isopropoxyphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
666890 Aldrich

6-Bromo-2-fluoro-3-isopropoxyphenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents,
InChI Key   JIDCDBHYZFYNSO-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Other Notes

Contains varying amounts of anhydride

Packaging

2, 10 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?