• USA Home
  • 667838 - Dodecanethiol functionalized silver nanoparticles

EMAIL THIS PAGE TO A FRIEND
667838 Aldrich

Dodecanethiol functionalized silver nanoparticles

3-7 nm particle size (DLS), 0.25 % (w/v) in hexane

Purchase

Properties

Related Categories 47: Ag, Biomedical Materials, Functionalized Silver Nanowires, Materials Science, Nanomaterials,
description   nanoparticles
form   solution
concentration   0.25 % (w/v) in hexane
refractive index   n20/D 1.3857
particle size   3-7 nm (DLS)
density   0.6652 g/mL at 25 °C
functional group   Thiol

Description

General description

Silver nanoparticles (AgNPs) are chemically modified with dodecanethiol to prevent the formation of aggregations. Functionalized silver nanoparticles are essentially used to reinforce polymer composites.

Application

Dispersion of AgNPs inhibits bond formation between fullerene and a bisporphyrin. Effect of AgNPs in decreasing the binding strength of a non-covalent system of pthalocyanine fulleropyrrolidine has been investigated.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 1208 3 / PGII
WGK Germany 
3
Flash Point(F) 
-18.4 °F
Flash Point(C) 
-28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?