EMAIL THIS PAGE TO A FRIEND
668052 Aldrich

Lithium fluoride

fused (pieces), 2-15 mm, 99.995%

Synonym: Fluorolithium

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Lithium, Lithium Fluoride (LiF),
InChI Key   PQXKHYXIUOZZFA-UHFFFAOYSA-M
assay   99.995%
form   fused (pieces)
particle size   2-15 mm
mp   845 °C(lit.)
density   2.64 g/mL at 25 °C(lit.)

Description

Packaging

10, 25, 100 g in poly bottle

Price and Availability


Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 3288 6.1 / PGIII
WGK Germany 
3
Protocols & Articles

Articles

Complex Hydrides: A New Category of Solid-state Lithium Fast-ion Conductors

Research and development of solid-state lithium fast-ion conductors is crucial because they can be potentially used as solid electrolytes in all-solid-state batteries, which may solve the safety and ...
Motoaki Matsuo,1 Hiroyuki Oguchi,1,2 Hideki Maekawa,2 Hitoshi Takamura,2Shin-ichi Orimo,1*
Material Matters Volume 5 Article 4
Keywords: Calorimetry, Deposition, Diffraction, Diffusion, Melting, Nuclear magnetic resonance spectroscopy, Raman spectroscopy, Search, Spectra, Spectroscopy, Substitutions, Support, X-Ray diffraction, transformation

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?