• USA Home
  • 668079 - Ethylenediaminetetraacetic acid ruthenium(III) chloride complex, monoamide, polymer-bound

EMAIL THIS PAGE TO A FRIEND
668079 Aldrich

Ethylenediaminetetraacetic acid ruthenium(III) chloride complex, monoamide, polymer-bound

50-100 mesh, extent of labeling: 0.1-0.5 mmol/g Ru loading, 1 % cross-linked

Synonym: EDTA ruthenium(III) chloride complex, monoamide, polymer-bound

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Ru Catalysts, Ruthenium Catalysts
crosslinking   1 % cross-linked
extent of labeling   0.1-0.5 mmol/g Ru loading
particle size   50-100 mesh

Description

Application

A mild catalyst for the Strecker reaction.

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Strecker reaction

The one-pot synthesis of a-amino nitriles by reaction of an aldehyde, ammonia, and hydrogen cyanide is commonly known as the Strecker reaction.1 Recent modifications to the traditional Strecker react...
Aldrich ChemFiles 2006, 6.7, 10.
Keywords: Chromatography, Column chromatography, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Strecker synthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?