EMAIL THIS PAGE TO A FRIEND
668575 Aldrich

QuadraPure® DET

450-650 μm, extent of labeling: 1.0-2.0 mmol/g loading, macroporous

Synonym: QuadraPure® Bis(ethyl mercaptan)

Purchase

Properties

Related Categories Chemical Synthesis, QuadraPure, Scavengers, Solid Supported Synthesis, Specialty Synthesis More...
form   macroporous
extent of labeling   1.0-2.0 mmol/g loading
particle size   450-650 μm

Description

Application

Metal Scavenger: Pd in acidic media and with phosphines, Co, Fe, Ni, Rh

Reaxa Technology

Packaging

25, 100 g in glass bottle

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Reaxa Technology

Easier, Faster, and Cleaner Reactions In partnership with Reaxa Ltd., Sigma-Aldrich is pleased to offer several technology platforms that allow for easier, faster, and cleaner processes. The technolo...
Keywords: Catalysis, Filtration, Metal scavengers, Methods, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?