• USA Home
  • 669423 - 2-Bromo-3′-chloro-4′-fluoroacetophenone

EMAIL THIS PAGE TO A FRIEND
669423 Aldrich

2-Bromo-3′-chloro-4′-fluoroacetophenone

96%

Synonym: 2-Bromo-1-(3-chloro-4-fluorophenyl)ethanone, 3′-Chloro-4′-fluorophenacyl bromide

  • CAS Number 63529-30-6

  • Empirical Formula (Hill Notation) C8H5BrClFO

  • Molecular Weight 251.48

  •  MDL number MFCD05154991

  •  PubChem Substance ID 24885047

Purchase

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?