Synonym: Polyethylene glycol 3400 monoacid
CAS Number 117786-94-4
Linear Formula HO(CH2CH2O)nCH2CH2COOH
MDL number MFCD09750435
| Related Categories | COOH Functionalized, COOH-PEG-Hydroxyl (COOH-PEG-OH), Chemical Synthesis, Heterobifunctional PEGs, Hydroxyl Functionalized, |
| storage temp. | −20°C |
100, 500 mg in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?