• USA Home
  • 671355 - Evonik Ligand Kit for CX Coupling Reactions

EMAIL THIS PAGE TO A FRIEND
671355 Aldrich

Evonik Ligand Kit for CX Coupling Reactions

Synonym: cataCXium® Ligand Toolbox

Purchase

Description

General description

sold in collaboration with Solvias AG

Legal Information

cataCXium is a registered trademark of Evonik Degussa GmbH

Protocols & Applications

Efficient Suzuki Coupling with cataCXium Ligands and Complexes

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

cataCXium® A, 95% SDS 671479
Di(1-adamantyl)-n-butylphosphine hydriodide, 95% SDS 671584
Di(1-adamantyl)benzylphosphine, 95% SDS 671800
trans-Bis(acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II), 98% SDS 671916
2-(Dicyclohexylphosphino)-1-phenyl-1H-pyrrole, 95% SDS 672017
2-(Dicyclohexylphosphino)-1-(2,4,6-trimethyl-phenyl)-1H-imidazole, 95% SDS 672122
2-(Dicyclohexylphosphino)-1-phenylindole, 95% (NMR) SDS 672246
2-(Di-tert-butylphosphino)-1-phenylindole, 95% SDS 672343
2-(Dicyclohexylphosphino)-1-(2-methoxyphenyl)-1H-pyrrole, 95% SDS 672459
2-(Di-tert-butylphosphino)-1-(2-methoxyphenyl)-1H-pyrrole, 95% SDS 672564
2-(Di-tert-butyl-phosphino)-1-phenyl-1H-pyrrole, 95% SDS 672661
(9-Butyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate, 97% SDS 672785
(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate, 95% SDS 691100
Dicyclohexyl[9-(3-phenylpropyl)-9-fluorenyl]phosphonium tetrafluoroborate, 95% SDS 688681
Dicyclohexyl-{2-sulfo-9-[3-(4-sulfo-phenyl)propyl]-9-fluorenyl}phosphonium-hydrogensulfate, 95% SDS 688800
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Palladacycle Coupling Catalysts

Palladacyclic catalysts developed by Bedford’s group are examples of a select group of catalysts capable of affecting a variety of coupling reactions using difficult to activate aryl chlorides at ver...
William Sommer
Aldrich ChemFiles 2007, 7.10, 17.
Keywords: Aminations, Catalysis, Chemfiles, Coupling reactions, Ligands, Protocols, Solvents, Stille coupling, Suzuki coupling

cataCXium® - Ligands and Complexes for Efficient Cross-Coupling Reactions

Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlor...
Keywords: Aminations, Buchwald-Hartwig amination, Cancer, Catalysis, Coupling reactions, Cross couplings, Industries, Ligands, Pharmaceutical, Polymer science, Suzuki coupling, transformation

cataCXium®

Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlor...
William Sommer
ChemFiles Volume 7 Article 10
Keywords: Aminations, Applications, Buchwald-Hartwig amination, Catalysis, Coupling reactions, Cross couplings, Industries, Ligands, Pharmaceutical, Polymer science, Suzuki coupling, transformation

Related Content

cataCXium® - Ligands and Complexes for Efficient Cross-Coupling Reactions

Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlor...
Keywords: Aminations, Applications, Buchwald-Hartwig amination, Cancer, Catalysis, Coupling reactions, Cross couplings, Industries, Ligands, Pharmaceutical, Polymer science, Suzuki coupling, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?