• USA Home
  • 672149 - 1-(4-Methoxyphenyl)-1-[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]-1-phenylmethyl chloride

EMAIL THIS PAGE TO A FRIEND
672149 Aldrich

1-(4-Methoxyphenyl)-1-[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]-1-phenylmethyl chloride

>90%

Synonym: 1-[Chloro[4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phenyl]phenylmethyl]-4-methoxy-benzene, F17 MMT Cl

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Building Blocks and Precursors, Specialty Synthesis
InChI Key   FSYYFUVFEYRUHN-UHFFFAOYSA-N
assay   >90%
mp   89-95 °C

Description

Packaging

1 g in poly bottle

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?