• USA Home
  • 672580 - 1,1-Di-(4-methoxyphenyl)-1-[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]methyl chloride

EMAIL THIS PAGE TO A FRIEND
672580 Aldrich

1,1-Di-(4-methoxyphenyl)-1-[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]methyl chloride

≥90%

Synonym: 1,1′-{Chloro[4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phenyl]methylene}-bis(4-methoxybenzene), F17 DMT Cl

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Reagents, Specialty Synthesis
InChI Key   BYTTZNJBGFBIDO-UHFFFAOYSA-N
assay   ≥90%
mp   131-137 °C

Description

Packaging

1 g in glass bottle

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?