• USA Home
  • 672696 - 1,1-Di-(4-methoxyphenyl)-1-[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]methanol

EMAIL THIS PAGE TO A FRIEND
672696 Aldrich

1,1-Di-(4-methoxyphenyl)-1-[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]methanol

97% (H-NMR)

Synonym: F17 DMT OH, [4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl)phenyl]-bis(4-methoxyphenyl)methanol

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Building Blocks and Precursors, Specialty Synthesis
InChI Key   WKTBNLMPYCYJER-UHFFFAOYSA-N
assay   97% (H-NMR)

Description

Packaging

1 g in poly bottle

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?