-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym:
Tris[4-
CAS Number 322647-82-5
Empirical Formula (Hill Notation) C39H18F39P
Molecular Weight 1258.47
MDL number MFCD06412555
PubChem Substance ID 24885137
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Fluorous Synthesis, Phosphine Ligands, Phosphorus Compounds, |
| assay | ≥90% |
Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?