• USA Home
  • 673641 - Vinylboronic acid 2-methyl-2,4-pentanediol ester

EMAIL THIS PAGE TO A FRIEND
673641 Aldrich

Vinylboronic acid 2-methyl-2,4-pentanediol ester

contains phenothiazine as stabilizer, 95%

Synonym: 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane, Vinylboronsäure-2-methyl-2,4-pentandiolester

Purchase

Properties

Related Categories Alkenyl Boronate Esters, Boronate Esters, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents More...
InChI Key   LFOAVJXWGLVDDI-UHFFFAOYSA-N
assay   95%
contains   phenothiazine as stabilizer
refractive index   n20/D 1.429
density   0.893 g/mL at 25 °C
storage temp.   2-8°C

Description

Other Notes

Purity excludes stabilizer

Stabilized with up to 1 wt% of Phenothiazine

Packaging

1, 10 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles

Articles

Boronic Acid Esters

The synthesis of biaryl compounds via the Suzuki–Miyaura coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura et al. demonst...
Chemfiles Volume 4 Article 2
Keywords: Coupling reactions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?