EMAIL THIS PAGE TO A FRIEND
67470 Aldrich

3-Methyl-1-hexene

technical, ≥85% (GC)

  • CAS Number 3404-61-3

  • Linear Formula CH3CH2CH2CH(CH3)CH=CH2

  • Molecular Weight 98.19

  •  Beilstein Registry Number 1733465

  •  EC Number 222-283-1

  •  MDL number MFCD00009407

  •  PubChem Substance ID 57652087

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   RITONZMLZWYPHW-UHFFFAOYSA-N
grade   technical
assay   ≥85% (GC)
impurities   ~10% 5-methyl-1-hexene
refractive index   n20/D 1.393
bp   83-84 °C
density   0.695 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3295BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?