• USA Home
  • 675377 - (R)-(−)-1-Amino-6-hydroxyindan

EMAIL THIS PAGE TO A FRIEND
675377 Aldrich

(R)-(−)-1-Amino-6-hydroxyindan

97%

  • CAS Number 169105-01-5

  • Empirical Formula (Hill Notation) C9H11NO

  • Molecular Weight 149.19

  •  MDL number MFCD04972514

Purchase

Properties

Related Categories Amino Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   MOUPGBDOLGDVNW-SECBINFHSA-N
assay   97%
mp   195-205 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?