-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym:
4-(1H,1H,2H,2H,3H,3H-
CAS Number 957206-65-4
Empirical Formula (Hill Notation) C16H13F13O2
Molecular Weight 484.25
MDL number MFCD04973816
PubChem Substance ID 24885390
| Related Categories | Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Protecting Groups, Specialty Synthesis |
| assay | ≥97.0% |
1 g in poly btl
Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?