• USA Home
  • 68182 - Potassium methyl sulfate

EMAIL THIS PAGE TO A FRIEND
68182 Aldrich

Potassium methyl sulfate

purum, ≥95.0% (T)

Synonym: Methyl sulfate potassium salt

  • CAS Number 562-54-9

  • Linear Formula CH3OSO3K

  • Molecular Weight 150.20

  •  Beilstein Registry Number 3710745

  •  EC Number 209-231-3

  •  MDL number MFCD00040472

  •  PubChem Substance ID 24885658

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Organic Sulfates/Sulfites, Sulfur Compounds More...
InChI Key   WBGWGHYJIFOATF-UHFFFAOYSA-M
grade   purum
assay   ≥95.0% (T)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?