EMAIL THIS PAGE TO A FRIEND
685674 Aldrich

(S)-Me-f-KetalPhos

Synonym: 1,1′-Bis[(3aS,4S,6S,6aS)-tetrahydro-2,2,4,6-tetramethyl-5H-phospholo[3,4-d]-1,3-dioxol-5-yl]ferrocene, 1,1-Bis[(2S,3S,4S,5S)-2,5-dimethyl-3,4-O-isopropylidene-3,4-dihydroxyphospholanyl]ferrocene

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Chiral Quest Ligands and Complexes, Privileged Ligands and Complexes More...
InChI Key   VMJRCJPLHRKPIW-DYCBUYNASA-N
mp   151.6-158.4 °C

Description

Legal Information

Sold in collaboration with Chiral Quest for research purposes only. US Patent No.: 6,337,406 and 6,727,377.

Application

Chiral Quest Phosphine Ligands for Asymmetric Hydrogenation

• Chiral ferrocenyl phosphine ligand used for catalysis of asymmetric hydrogenation reaction

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

ChiralQuest Phosphine Ligands and Complexes

Professor Xumu Zhang at Penn State has made remarkable advances by creating a toolbox of chiral phosphines which can be used on a variety of substrates, some of which have been historically resistant...
William Sommer and Daniel Weibel
Aldrich ChemFiles 2008, 8.2, 47.
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Chemfiles, Cyclizations, Hydrogenations, Ligands, Reductions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?