• USA Home
  • 688061 - N1,N4-Bis(4-butylphenyl)benzene-1,4-diamine

EMAIL THIS PAGE TO A FRIEND
688061 Aldrich

N1,N4-Bis(4-butylphenyl)benzene-1,4-diamine

97%

Synonym: N,N′-Bis(4-butylphenyl)benzene-1,4-diamine

Purchase

Properties

Related Categories Materials Science, Organic and Printed Electronics, Synthetic Intermediates, Synthetic Tools and Reagents
InChI Key   XFNDFNOXZWWULM-UHFFFAOYSA-N
assay   97%
mp   125-129 °C

Description

Application

Monomer for synthesis of charge transport and light emitting polymers.

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Development of Organic Semiconductors from Highly Ordered Oligo and Polythiophenes

Nicholas S. Colella, Lei Zhang, Alejandro L. Briseño* Polymer Science & Engineering Department, University of Massachusetts, Amherst, Massachusetts 01003 *Email: abriseno@mail.pse.umass.edu
Keywords: Absorption, Building blocks, Catalysis, Coupling reactions, Cross couplings, Crystallization, Electronics, Environmental, Grignard Reaction, Metathesis, Nanomaterials, Nucleic acid annealing, Organic electronics, Photovoltaics, Polymer science, Polymerization reactions, Semiconductor, Solar cells, Ultraviolet-Visible spectroscopy

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?