• USA Home
  • 688924 - cis-2-(Aminomethyl)cyclohexanol hydrochloride

EMAIL THIS PAGE TO A FRIEND
688924 Aldrich

cis-2-(Aminomethyl)cyclohexanol hydrochloride

  • CAS Number 24947-68-0

  • Empirical Formula (Hill Notation) C7H15NO · HCl

  • Molecular Weight 165.66

  •  MDL number MFCD09260355

Purchase

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?