• USA Home
  • 691232 - (S)-(−)-α-Propargylalanine

EMAIL THIS PAGE TO A FRIEND
691232 Aldrich

(S)-(−)-α-Propargylalanine

≥98.0% (HPLC)

Synonym: (S)-2-Amino-2-methyl-4-pentynoic acid

  • Empirical Formula (Hill Notation) C6H9NO2

  • Molecular Weight 127.14

  •  MDL number MFCD10567140

  •  PubChem Substance ID 329761395

Purchase

Properties

Related Categories Amines, Building Blocks, C6, Carbonyl Compounds, Carboxylic Acids,
InChI Key   FSBNDYYRTZBHAN-LURJTMIESA-N
assay   ≥98.0% (HPLC)
optical purity   ee: ≥98.0% (HPLC)
impurities   ≤15% water

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?