• USA Home
  • 692697 - (R,R,R)-1-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)-2,5-diphenylpyrrolidine

EMAIL THIS PAGE TO A FRIEND
692697 Aldrich

(R,R,R)-1-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)-2,5-diphenylpyrrolidine

90%

Synonym: 1-(11bR)-Dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-yl-(2R,5R)-2,5-diphenylpyrrolidine

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, DSM MonoPhos Phosphoramidites, Privileged Ligands and Complexes More...
InChI Key   SQEYSQAZYNQDSV-ROJLCIKYSA-N
assay   90%
optical activity   [α]22/D −237°, c = 0.5 in chloroform
mp   191-196 °C
storage temp.   −20°C

Description

Application

Used in asymmetric hydrogenation.

DSM MonoPhos Family of Highly Efficient Privileged Ligands

Packaging

100, 500 mg in glass insert

Legal Information

Sold under license from DSM for research purposes only.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?