EMAIL THIS PAGE TO A FRIEND
693669 Aldrich

Ytterbium

foil, thickness 1 mm, 99.9% trace rare earth metals basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Metals,
InChI Key   NAWDYIZEMPQZHO-UHFFFAOYSA-N
assay   99.9% trace rare earth metals basis
form   foil
resistivity   28 μΩ-cm, 20°C
thickness   1 mm
bp   1196 °C(lit.)
mp   824 °C(lit.)
density   6.54 g/mL at 25 °C(lit.)

Description

Application

Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

Packaging

1 ea in rigid mailer

Quantity

1 ea = 25 x 25 mm

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ZG1925000
Protocols & Articles

Articles

Functional Magnetic Materials: Synergy Between Basic Science and Evolving Technology

S. B. Roy Materials and Advanced Accelerator Sciences Division Raja Ramanna Centre for Advanced Technology Indore-452013, India Email: sbroy@rrcat.gov.in
Keywords: Crystallization, Microscopy, Phase transitions, Reductions, transformation

Kinetic Energy Harvesting by Magnetostrictive Materials

Daniele Davino Department of Engineering University of Sannio, 82100 Benevento, Italy Email: davino@unisannio.it
Keywords: Electronics, Environmental

Metal Hydrides for NiMH Battery Applications

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications. The Partnership for a New Generation of V...
Dhanesh Chandra,* Wen-Ming Chien and Anjali Talekar
Material Matters Volume 6 Article 2
Keywords: Absorption, Applications, Automotive, Capillary electrophoresis, Combustion, Degradations, Electrochemical analysis, Electronics, Environmental, Help, Ion-exchange chromatography, Phase transitions, Precipitation, Purification, Separation, Substitutions, Type

Responsive Rare-Earth Materials and their Applications

Yaroslav Mudryk Ames Laboratory of U.S. Department of Energy, Iowa State University Ames, Iowa 50011 USA Email: slavkomk@ameslab.gov
Yaroslav Mudryk
Material Matters, 2016, 11.4
Keywords: Diffraction, PAGE, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?