• USA Home
  • 697680 - (S)-2-[(Boc-amino)methyl]-3-phenylpropionic acid

EMAIL THIS PAGE TO A FRIEND
697680 Aldrich

(S)-2-[(Boc-amino)methyl]-3-phenylpropionic acid

97%

Synonym: (S)-N-Boc-2-benzyl-β-alanine, (S)-2-Benzyl-3-(Boc-amino)propionic acid

Purchase

Properties

Related Categories Asymmetric Synthesis, Beta-Amino Acids, Carboxylic Acids, Chemical Biology, Chemical Synthesis,
InChI Key   ZYCITKXROAFBAR-LBPRGKRZSA-N
assay   97%
optical activity   [α]22/D −15.0°, c = 1 in chloroform
mp   92-100 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?