• USA Home
  • 69900 - 1-Morpholinocyclododecene

EMAIL THIS PAGE TO A FRIEND
69900 Aldrich

1-Morpholinocyclododecene

≥95.0%

Synonym: 4-(1-Cyclododecen-1-yl)morpholine

  • CAS Number 3725-39-1

  • Empirical Formula (Hill Notation) C16H29NO

  • Molecular Weight 251.41

  •  Beilstein Registry Number 611459

  •  EC Number 223-080-0

  •  MDL number MFCD00042788

  •  PubChem Substance ID 57652224

Purchase

Properties

Related Categories Building Blocks, C11 to C20+, Chemical Synthesis, Heterocyclic Building Blocks, Morpholines/Thiomorpholines More...
InChI Key   SZTBSJOMNRSBGA-MHWRWJLKSA-N
assay   ≥95.0%
refractive index   n20/D 1.516
bp   129-131 °C/0.05 mmHg(lit.)
density   0.988 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Versatile enamine, used e.g. for the synthesis of long-chain fatty acids by chain elongation; Synthesis of meta-cyclophane

Packaging

10 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?