• USA Home
  • 699365 - 1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
699365 Aldrich

1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carboxaldehyde

  • CAS Number 957483-88-4

  • Empirical Formula (Hill Notation) C9H12N2O2

  • Molecular Weight 180.20

  •  MDL number MFCD07186538

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C9, Carbonyl Compounds, Chemical Synthesis,
InChI Key   LJKATJHBJPFKBQ-UHFFFAOYSA-N
refractive index   n20/D 1.531
density   1.126 g/mL at 25 °C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?