• USA Home
  • 69943 - Morpholinomethyl-polystyrene

EMAIL THIS PAGE TO A FRIEND
69943 Aldrich

Morpholinomethyl-polystyrene

extent of labeling: ~3.5 mmol/g base loading

Synonym: Copolymer of styrene and divinylbenzene, morpholinomethylated

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Solid Supported Synthesis,
extent of labeling   ~3.5 mmol/g base loading
impurities   ≤0.3 mmol/g Cl (resin)
matrix   crosslinked with 2% DVB
particle size   200-400 mesh

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?