• USA Home
  • 700525 - N,N′-Bis(p-toluenesulfonyl)hydrazine

EMAIL THIS PAGE TO A FRIEND
700525 Aldrich

N,N′-Bis(p-toluenesulfonyl)hydrazine

97%

Synonym: 1,2-Bis(p-tolylsulfonyl)hydrazine, 4-Methylbenzenesulfonic acid 2-[(4-methylphenyl)sulfonyl]hydrazide, N,N′-Ditosylhydrazine

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   CVRIWLRSJCDOOX-UHFFFAOYSA-N
assay   97%
mp   220-224 °C
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?