• USA Home
  • 703303 - (5,6,7,8-Tetrahydro-2-naphthoyl)acetonitrile

EMAIL THIS PAGE TO A FRIEND
703303 Aldrich

(5,6,7,8-Tetrahydro-2-naphthoyl)acetonitrile

97%

Synonym: 3-(1,2,3,4-Tetrahydronaphthalen-6-yl)-3-oxopropanenitrile, 3-(5,6,7,8-Tetrahydro-2-naphthyl)-3-oxopropionitrile

Purchase

Properties

Related Categories Building Blocks, C10 to C27, C13 to C14, Carbonyl Compounds, Chemical Synthesis,
InChI Key   MMCQHIGQRVUVMS-UHFFFAOYSA-N
assay   97%
mp   93-100 °C

Description

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?