• USA Home
  • 703605 - Deprotection kit I for N-O debenzylation
EMAIL THIS PAGE TO A FRIEND
703605 Aldrich
Deprotection kit I for N-O debenzylation
Purchase
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
Legal Information
Sold in collaboration with Johnson Matthey Catalyst for research purposes only.
Price and Availability
Cross-Coupling Guide
Safety & Documentation
Safety Information
RIDADR 
UN 3089 4.1/PG 2
Documents
Certificate of Analysis
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
What is Product 703605, Deprotection Kit I for N-O debenzylation, typically used for?
Per the supplier for product 703605, it has been developed with a range of more active and selective catalysts with reduced metal loadings designed for the O-debenzylation of benzyl protected alcohols and acids, N-debenzylation of amines and amides and Cbz (carbamate) type protection of amines.
What is the quantity of each catalyst in Product 703605, Deprotection Kit I for N-O debenzylation?
The kit is supplied with 5g of each catalyst.
What are the solvents used for N-debenzylation when using Product 703605, Deprotection Kit I for N-O debenzylation?
Commonly used solvents for N-debenzylation are Alcohols (+H+), ethyl acetate, and acetic acid.
What are the common solvents for O-debenzylation when using Product 703605, Deprotection Kit I for N-O debenzylation?
Commonly used solvents for O-debenzylation are THF, ethanol, ethyl acetate, and acetic acid. Reaction rates were fastest in THF, slightly slower in ethanol and ethyl acetate, and slowest in acetic acid. For less active catalysts, ethanol was a poor solvent choice due to limited substrate solubility.
What are the recommended reaction conditions when using Product 703605, Deprotection Kit I for N-O debenzylation?  
N-Debenzylation reactions occur best at low pressures (1-10 bar), at a temperature between 30-100ºC where the increase of temperature from 50°C to 70°C increases the rate by a factor of 3. The recommendations for N-debenzylation have a pressure of 1-10 bar and a temperature range of 25-50ºC. However, a variety of solvents, temperatures, pressures, and catalyst loadings should be evaluated to arrive at an optimized set of reaction conditions.
Show more questions
Protocols & Articles
Articles
Catalytic Deprotection—DEPRO™ Catalyst Kit/Catalytic Deprotection
In the manufacture of pharmaceuticals and fine chemicals there is often a requirement for a protection strategy to minimize possible side reactions during a synthesis. Small, easily removed protectin...
Robert J. McNair
Aldrich ChemFiles 2009, 9.2, 21.
Keywords: Catalysis, Gas chromatography, High performance liquid chromatography, Hydrogenations, Oxidations, Pharmaceutical, Solvents, Support, Type
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?