EMAIL THIS PAGE TO A FRIEND
703613 Aldrich

PEG dibromide

average mol wt 3,480

Synonym: Polyethylene glycol dibromide

Purchase

Properties

Related Categories Bromide, Homobifunctional PEGs, Materials Science, Poly(ethylene glycol) and Poly(ethylene oxide), Polymer Science,
mol wt   average mol wt 3,480
mp   56-65 °C

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?