EMAIL THIS PAGE TO A FRIEND
704458 Aldrich

PEG-di-p-tosylate

average Mn 1,800

Purchase

Properties

Related Categories Homobifunctional PEGs, Materials Science, Poly(ethylene glycol) and Poly(ethylene oxide), Polymer Science, Polymers,
mol wt   average Mn 1,800
mp   26-34 °C
  43-47 °C
Ω-end   Tosylate
α-end   Tosylate

Description

Packaging

5 g in glass bottle

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?