• USA Home
  • 705101 - Barbituric acid-13C4,15N2

EMAIL THIS PAGE TO A FRIEND
705101 Aldrich

Barbituric acid-13C4,15N2

≥98 atom %, ≥97% (CP)

Synonym: 2,4,6-Trihydroxypyrimidine-13C4,15N2

  • Empirical Formula (Hill Notation) 13C4H415N2O3

  • Molecular Weight 134.04

  •  MDL number MFCD11656097

  •  PubChem Substance ID 329762504

Purchase

Properties

Related Categories Metabolites and Bioactive Compounds, Other Bioactive Compounds, Stable Isotopes More...
InChI Key   HNYOPLTXPVRDBG-IDEBNGHGSA-N
isotopic purity   ≥98 atom %
assay   ≥97% (CP)
mp   248-252 °C ((dec.))
mass shift   M+6
storage temp.   −20°C

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?