Synonym: MDA
CAS Number 82001-53-4
Empirical Formula (Hill Notation) C12H24O2S
Molecular Weight 232.38
MDL number MFCD08705252
| Related Categories | Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis, |
| assay | 99% |
| mp | 45-50 °C |
| storage temp. | 2-8°C |
250 mg in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?