• USA Home
  • 70694 - (±)-1-(1-Naphthyl)ethanol

EMAIL THIS PAGE TO A FRIEND
70694 Aldrich

(±)-1-(1-Naphthyl)ethanol

≥99.0% (GC)

Synonym: (±)-α-Methyl-1-naphthalenemethanol

  • CAS Number 1517-72-2

  • Empirical Formula (Hill Notation) C12H12O

  • Molecular Weight 172.22

  •  Beilstein Registry Number 1865349

  •  EC Number 216-172-7

  •  MDL number MFCD00016806

  •  PubChem Substance ID 57652257

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   CDRQOYRPWJULJN-UHFFFAOYSA-N
assay   ≥99.0% (GC)
mp   63-65 °C

Description

Packaging

10 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?