• USA Home
  • 707775 - Guanosine-15N5 5′-triphosphate sodium salt solution

EMAIL THIS PAGE TO A FRIEND
707775 Aldrich

Guanosine-15N5 5′-triphosphate sodium salt solution

98 atom % 15N, 95% (CP), 100 mM in 5 mM Tris HCl

Synonym: GTP-15N5 sodium salt solution

  • Empirical Formula (Hill Notation) C10H1615N5O14P3 · xNa+

  • Molecular Weight 528.15 (free acid basis)

Purchase

Properties

Related Categories Alphabetical Listings, G-H, Nucleotides, Ribonucleotide triphosphates, Stable Isotopes More...
isotopic purity   98 atom % 15N
assay   95% (CP)
contains   5 mM Tris buffer
concentration   100 mM in 5 mM Tris HCl
  100 mM in H2O
mass shift   M+5

Description

Other Notes

Molecular weight and empirical formula based on the free acid.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

High Quality Products for Nucleic Acid Research

“Over the course of several years now, my group hasconsistently been happy with the quality of the NTPsand services provided by ISOTEC. We are particularlypleased by the fact that the NTPs are often ...
Keywords: High performance liquid chromatography, Nuclear magnetic resonance spectroscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?